Assay Details
| Gene Symbol: Zfp799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177359.5 | NP_796333.3 | 2 - 3 | 293 | 123 | |
| GenBank mRNA | AK031686.1 | - | 2 - 3 | 281 | 123 | |
| AK083640.1 | - | 2 - 3 | 270 | 123 | ||
| AK137037.1 | - | 2 - 3 | 270 | 123 | ||
| BC096552.1 | - | 2 - 3 | 243 | 30618827 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp799
Genomic Map