Assay Details
| Gene Symbol: Brd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286630.1 | NP_001273559.1 | 4 - 5 | 733 | 142 | |
| NM_020508.4 | NP_065254.3 | 4 - 5 | 689 | 142 | ||
| NM_198094.2 | NP_932762.2 | 4 - 5 | 679 | 142 | ||
| GenBank mRNA | AF273217.1 | - | 3 - 4 | 604 | 142 | |
| AF461395.1 | - | 4 - 5 | 665 | 142 | ||
| AF461396.1 | - | 4 - 5 | 665 | 142 | ||
| AK147545.1 | - | 4 - 5 | 698 | 142 | ||
| AK154514.1 | - | 3 - 4 | 664 | 142 | ||
| BC011541.1 | - | 3 - 4 | 525 | 142 | ||
| BC013666.1 | - | 3 - 4 | 662 | 142 | ||
| BC082782.1 | - | 3 - 4 | 477 | 142 | ||
| BC103773.1 | - | 3 - 4 | 603 | 142 | ||
| BC138834.1 | - | 4 - 5 | 659 | 8861856 | 142 | |
| BC138835.1 | - | 4 - 5 | 658 | 8861857 | 142 | |
More Information
Updated Mapping Information
Set Membership: |
|

Brd4
Genomic Map