Assay Details
| Gene Symbol: Lypd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083884.1 | NP_001077353.1 | 3 - 4 | 233 | 83 | |
| NM_027339.2 | NP_081615.2 | 2 - 3 | 168 | 83 | ||
| GenBank mRNA | AK008940.1 | - | 3 - 4 | 222 | 83 | |
| BC026828.1 | - | 2 - 3 | 143 | 4218333 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lypd8
Genomic Map