Assay Details
| Gene Symbol: Lypd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083884.1 | NP_001077353.1 | 5 - 6 | 508 | 94 | |
| NM_027339.2 | NP_081615.2 | 4 - 5 | 443 | 94 | ||
| GenBank mRNA | AK008940.1 | - | 5 - 6 | 497 | 94 | |
| AK136583.1 | - | 434 | 94 | |||
| BC026828.1 | - | 4 - 5 | 418 | 4218333 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lypd8
Genomic Map