Assay Details
| Gene Symbol: Lypd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083884.1 | NP_001077353.1 | 6 - 7 | 645 | 98 | |
| NM_027339.2 | NP_081615.2 | 5 - 6 | 580 | 98 | ||
| GenBank mRNA | AK008940.1 | - | 6 - 7 | 634 | 98 | |
| AK136583.1 | - | 571 | 98 | |||
| BC026828.1 | - | 5 - 6 | 555 | 4218333 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670766 | AK136583.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670766 | BC026828.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670766 | NM_001083884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670766 | NM_027339.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670766 | AK008940.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670766 | AK136583.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670766 | BC026828.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670766 | NM_001083884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670766 | NM_027339.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670766 | AK008940.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136676 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416606 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 63623 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593611 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | AK005370.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | AK042930.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | AK088768.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | AK129452.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | AK145459.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | AK146670.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | AK159009.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | AK159525.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | AK159930.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | AK214151.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | BC014855.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | BC034011.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | BC052862.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | BC066213.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | BC071200.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | NM_011241.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924321 | U08110.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 294927 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106140 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 353697 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 59173 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138744 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103438 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670766 | AK136583.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670766 | BC026828.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670766 | NM_001083884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670766 | NM_027339.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670766 | AK008940.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670766 | AK136583.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670766 | BC026828.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670766 | NM_001083884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670766 | NM_027339.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670766 | AK008940.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830670 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138744 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103438 | - | - |
| Whole Mouse Genome G4122A | A_51_P229403 | AK136583.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P229403 | AW121391.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P229403 | BC026828.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P229403 | NM_001083884.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P229403 | NM_027339.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670766 | AK136583.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670766 | BC026828.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670766 | NM_001083884.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670766 | NM_027339.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670766 | AK008940.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670766 | AK136583.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670766 | BC026828.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670766 | NM_001083884.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670766 | NM_027339.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670766 | AK008940.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103438 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138744 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09498630 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09395818 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09498632 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09498631 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09074723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09258050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09625947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09702396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09477584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09485217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09332049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09498630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09021170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830739 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10376371 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376373 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376369 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376368 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376367 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376370 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376372 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451441_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10376366 | - | - |
| Mouse Genome 430 2.0 Array | 1451441_at | - | - |
| Mouse Genome 430 2.0 Array | 1451441_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70163_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70163_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100005549.mm.1 | NM_001083884.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005549.mm.1 | NM_001083884.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005549.mm.1 | NM_027339.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005549.mm.1 | NM_027339.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005549.mm.1 | TC1100000689.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451441_at | - | - |
| Mouse Expression Set 430 | 1451441_PM_at | - | - |
| Murine Genome U74v2 Set | 94914_PM_at | - | - |
| Murine Genome U74v2 Set | 94914_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451441_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451441_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451441_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451441_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 487473 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1940541 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1227061 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227061 | - | - |

Lypd8
Genomic Map