Assay Details
| Gene Symbol: Nt5c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252374.1 | NP_001239303.1 | 7 - 8 | 495 | 99 | |
| NM_026004.3 | NP_080280.3 | 6 - 7 | 652 | 99 | ||
| GenBank mRNA | AK011894.1 | - | 6 - 7 | 550 | 99 | |
| BC038029.1 | - | 7 - 8 | 488 | 4018956 | 99 | |
| CB571852.1 | - | 7 - 8 | 500 | 99 | ||
| CX734167.1 | - | 2 - 3 | 99 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Nt5c3
Genomic Map