Assay Details
| Gene Symbol: Ubash3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177823.4 | NP_808491.2 | 11 - 12 | 1571 | 101 | |
| GenBank mRNA | AK040572.1 | - | 11 - 12 | 1532 | 101 | |
| AK041690.1 | - | 11 - 12 | 1538 | 101 | ||
| AK049106.1 | - | 10 - 11 | 1471 | 101 | ||
| AK169696.1 | - | 11 - 12 | 1526 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ubash3a
Genomic Map