Assay Details
| Gene Symbol: LOC102631757 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110503.1 | 1 - 2 | 222 | 65 | ||
| GenBank mRNA | AK050258.1 | - | 1 - 2 | 180 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102631757| Gene Symbol: LOC102631757 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110503.1 | 1 - 2 | 222 | 65 | ||
| GenBank mRNA | AK050258.1 | - | 1 - 2 | 180 | 65 | |
Set Membership: |
|