Assay Details
| Gene Symbol: Tcaim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013405.2 | NP_001013423.2 | 8 - 9 | 1073 | 72 | |
| GenBank mRNA | BC080690.1 | - | 8 - 9 | 1032 | 30111134 | 72 |
More Information
Updated Mapping Information
Set Membership: |
|

Tcaim
Genomic Map