Assay Details
| Gene Symbol: Shroom2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290684.1 | NP_001277613.1 | 2 - 3 | 349 | 97 | |
| NM_172441.3 | NP_766029.2 | 2 - 3 | 349 | 97 | ||
| GenBank mRNA | BF463332.1 | - | 2 - 3 | 186 | 97 | |
| DQ358971.1 | - | 2 - 3 | 330 | 97 | ||
| EF071946.1 | - | 2 - 3 | 349 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shroom2
Genomic Map