Assay Details
| Gene Symbol: Exog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172136.1 | NP_001165607.1 | 3 - 4 | 413 | 79 | |
| NM_172456.3 | NP_766044.1 | 2 - 3 | 378 | 79 | ||
| GenBank mRNA | AK028887.1 | - | 2 - 3 | 342 | 79 | |
| AK034654.1 | - | 3 - 4 | 375 | 79 | ||
| AK051300.1 | - | 3 - 4 | 377 | 79 | ||
| AK148426.1 | - | 2 - 3 | 359 | 79 | ||
| AK162566.1 | - | 2 - 3 | 341 | 79 | ||
| AK163555.1 | - | 2 - 3 | 342 | 79 | ||
| BC059920.1 | - | 3 - 4 | 356 | 79 | ||
| BC064730.1 | - | 3 - 4 | 428 | 79 | ||
| BC119024.1 | - | 2 - 3 | 342 | 8733773 | 79 | |
| BC144729.1 | - | 3 - 4 | 377 | 79 | ||
More Information
Set Membership: |
|

Exog
Genomic Map