Assay Details
| Gene Symbol: Exog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172136.1 | NP_001165607.1 | 4 - 5 | 555 | 108 | |
| NM_172456.3 | NP_766044.1 | 3 - 4 | 520 | 108 | ||
| GenBank mRNA | AK028887.1 | - | 3 - 4 | 484 | 108 | |
| AK034654.1 | - | 4 - 5 | 517 | 108 | ||
| AK051300.1 | - | 4 - 5 | 519 | 108 | ||
| AK148426.1 | - | 3 - 4 | 501 | 108 | ||
| AK162566.1 | - | 3 - 4 | 483 | 108 | ||
| AK163555.1 | - | 3 - 4 | 484 | 108 | ||
| BC059920.1 | - | 4 - 5 | 498 | 108 | ||
| BC064730.1 | - | 4 - 5 | 570 | 108 | ||
| BC119024.1 | - | 3 - 4 | 484 | 8733773 | 108 | |
| BC144729.1 | - | 4 - 5 | 519 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Exog
Genomic Map