| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900013212.mm.1 |
NM_001172136.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013212.mm.1 |
NM_001172136.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013212.mm.1 |
TC0900001582.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900013209.mm.1 |
NM_001172136.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013209.mm.1 |
NM_001172136.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013209.mm.1 |
NM_172456.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013209.mm.1 |
NM_172456.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013209.mm.1 |
TC0900001582.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900013211.mm.1 |
NM_001172136.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013211.mm.1 |
NM_001172136.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013211.mm.1 |
NM_172456.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013211.mm.1 |
NM_172456.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900013211.mm.1 |
TC0900001582.mm.1 |
MTA TC |