Assay Details
| Gene Symbol: Exog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172136.1 | NP_001165607.1 | 2 - 3 | 260 | 78 | |
| GenBank mRNA | AK034654.1 | - | 2 - 3 | 222 | 78 | |
| AK051300.1 | - | 2 - 3 | 224 | 78 | ||
| BC059920.1 | - | 2 - 3 | 203 | 78 | ||
| BC144729.1 | - | 2 - 3 | 224 | 78 | ||
More Information
Set Membership: |
|

Exog
Genomic Map