Assay Details
| Gene Symbol: Azi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048146.2 | NP_001041611.1 | 4 - 5 | 882 | 103 | |
| NM_001286508.1 | NP_001273437.1 | 4 - 5 | 800 | 103 | ||
| NM_013727.4 | NP_038755.1 | 4 - 5 | 882 | 103 | ||
| GenBank mRNA | AB007141.1 | - | 4 - 5 | 718 | 103 | |
| AK004992.1 | - | 4 - 5 | 861 | 103 | ||
| AK015942.1 | - | 4 - 5 | 703 | 103 | ||
| AK030076.1 | - | 4 - 5 | 714 | 103 | ||
| AK134714.1 | - | 4 - 5 | 785 | 103 | ||
| AK153477.1 | - | 4 - 5 | 846 | 103 | ||
| AK165549.1 | - | 4 - 5 | 768 | 103 | ||
| AK167783.1 | - | 4 - 5 | 826 | 103 | ||
| BC006851.1 | - | 4 - 5 | 777 | 3156857 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Azi2
Genomic Map