Assay Details
| Gene Symbol: Ly86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010745.2 | NP_034875.1 | 2 - 3 | 313 | 150 | |
| GenBank mRNA | AB007599.1 | - | 2 - 3 | 313 | 150 | |
| AK027929.1 | - | 2 - 3 | 302 | 150 | ||
| BC065783.1 | - | 2 - 3 | 218 | 150 | ||
| BC096414.1 | - | 2 - 3 | 281 | 849991 | 150 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ly86
Genomic Map