Assay Details
| Gene Symbol: Ly86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010745.2 | NP_034875.1 | 3 - 4 | 439 | 65 | |
| GenBank mRNA | AB007599.1 | - | 3 - 4 | 439 | 65 | |
| AK027929.1 | - | 3 - 4 | 428 | 65 | ||
| BC065783.1 | - | 3 - 4 | 344 | 65 | ||
| BC096414.1 | - | 3 - 4 | 407 | 849991 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ly86
Genomic Map