Assay Details
| Gene Symbol: Clcnkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019701.2 | NP_062675.2 | 1 - 2 | 41 | 54 | |
| GenBank mRNA | AF124847.1 | - | 1 - 2 | 34 | 54 | |
| AK146876.1 | - | 1 - 2 | 30 | 54 | ||
| AK165340.1 | - | 1 - 2 | 29 | 54 | ||
| BC019983.1 | - | 1 - 2 | 67 | 4188062 | 54 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF124847.1, BC019983.1, NM_019701.2 transcripts.
Set Membership: |
|

Clcnkb
Genomic Map