Assay Details
| Gene Symbol: Clcnkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019701.2 | NP_062675.2 | 5 - 6 | 545 | 74 | |
| GenBank mRNA | AK146876.1 | - | 5 - 6 | 534 | 74 | |
More Information
Set Membership: |
|

Clcnkb
Genomic Map