Assay Details
| Gene Symbol: Arhgef19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172520.2 | NP_766108.1 | 1 - 2 | 125 | 78 | |
| GenBank mRNA | AK031443.1 | - | 1 - 2 | 126 | 78 | |
| AK053075.1 | - | 1 - 2 | 126 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK053075.1, NM_172520.2 transcripts.
Set Membership: |
|

Arhgef19
Genomic Map