Assay Details
| Gene Symbol: Pla2g2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011109.2 | NP_035239.1 | 2 - 3 | 259 | 66 | |
| GenBank mRNA | AF112983.1 | - | 2 - 3 | 215 | 66 | |
| AF124374.1 | - | 2 - 3 | 189 | 66 | ||
| AF169407.1 | - | 2 - 3 | 259 | 66 | ||
| AF169408.1 | - | 2 - 3 | 259 | 66 | ||
| AK004232.1 | - | 2 - 3 | 220 | 66 | ||
| AK018005.1 | - | 2 - 3 | 216 | 66 | ||
| AK157392.1 | - | 2 - 3 | 216 | 66 | ||
| AK158364.1 | - | 2 - 3 | 187 | 66 | ||
| AK171797.1 | - | 2 - 3 | 219 | 66 | ||
| BC111806.1 | - | 2 - 3 | 248 | 40039182 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g2d
Genomic Map