| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400014475.mm.1 |
NM_175438.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014475.mm.1 |
NM_175438.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014475.mm.1 |
TC0400004196.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400014470.mm.1 |
NM_175438.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014470.mm.1 |
NM_175438.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014470.mm.1 |
TC0400004196.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400035179.mm.1 |
NM_175438.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400035179.mm.1 |
NM_175438.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400035179.mm.1 |
TC0400004196.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400014471.mm.1 |
NM_175438.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014471.mm.1 |
NM_175438.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014471.mm.1 |
TC0400004196.mm.1 |
MTA TC |