Assay Details
| Gene Symbol: Aldh4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175438.4 | NP_780647.3 | 5 - 6 | 700 | 70 | |
| GenBank mRNA | AK044712.1 | - | 5 - 6 | 496 | 70 | |
| BC024133.1 | - | 4 - 5 | 418 | 70 | ||
| BC027140.1 | - | 1 - 2 | 112 | 70 | ||
| BC039281.1 | - | 5 - 6 | 484 | 4975491 | 70 | |
| BC056226.1 | - | 5 - 6 | 584 | 5064715 | 70 | |
| BC060650.1 | - | 5 - 6 | 700 | 30354228 | 70 | |
| CF536864.1 | - | 1 - 2 | 62 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aldh4a1
Genomic Map