Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800017297.mm.1 |
NM_173876.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017297.mm.1 |
NM_173876.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017297.mm.1 |
TC0800002290.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017299.mm.1 |
NM_173876.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017299.mm.1 |
NM_173876.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017299.mm.1 |
TC0800002290.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017293.mm.1 |
NM_007711.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017293.mm.1 |
NM_007711.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017293.mm.1 |
NM_173873.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017293.mm.1 |
NM_173873.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017293.mm.1 |
NM_173874.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017293.mm.1 |
NM_173874.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017293.mm.1 |
NM_173876.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017293.mm.1 |
NM_173876.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017293.mm.1 |
TC0800002290.mm.1 |
MTA TC |