Assay Details
| Gene Symbol: Mfap3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027756.5 | NP_082032.1 | 1 - 2 | 225 | 127 | |
| GenBank mRNA | BC095950.1 | - | 1 - 2 | 171 | 5025281 | 127 |
| CJ111589.1 | - | 1 - 2 | 192 | 127 | ||
| CJ132157.1 | - | 1 - 2 | 175 | 127 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC095950.1, NM_027756.5 transcripts.
Set Membership: |
|

Mfap3l
Genomic Map