Assay Details
| Gene Symbol: Clcnka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146307.1 | NP_001139779.1 | 15 - 16 | 1813 | 79 | |
| NM_024412.3 | NP_077723.3 | 16 - 17 | 1817 | 79 | ||
| GenBank mRNA | AF124848.1 | - | 15 - 16 | 1910 | 79 | |
| AY373832.1 | - | 15 - 16 | 1768 | 79 | ||
| BC037077.1 | - | 16 - 17 | 1821 | 5042736 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clcnka
Genomic Map