Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400032301.mm.1 |
TC0400003892.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032301.mm.1 |
NM_172338.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032301.mm.1 |
NM_172338.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032300.mm.1 |
TC0400003892.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032300.mm.1 |
NM_172338.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032300.mm.1 |
NM_172338.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032302.mm.1 |
TC0400003892.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032302.mm.1 |
NM_172338.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032302.mm.1 |
NM_172338.2 |
Refseq |