Assay Details
| Gene Symbol: Agmat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081408.1 | NP_001074877.1 | 1 - 2 | 430 | 131 | |
| GenBank mRNA | AK017155.1 | - | 1 - 2 | 329 | 131 | |
| BC115693.2 | - | 1 - 2 | 327 | 40104399 | 131 | |
More Information
Set Membership: |
|

Agmat
Genomic Map