Assay Details
| Gene Symbol: Anxa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136089.2 | NP_001129561.1 | 2 - 3 | 264 | 62 | |
| NM_011922.3 | NP_036052.2 | 2 - 3 | 264 | 62 | ||
| GenBank mRNA | AK008808.1 | - | 2 - 3 | 188 | 62 | |
| AK020288.1 | - | 2 - 3 | 189 | 62 | ||
| AK131880.1 | - | 2 - 3 | 184 | 62 | ||
| AK168715.1 | - | 2 - 3 | 168 | 62 | ||
| BC125319.1 | - | 2 - 3 | 136 | 40129834 | 62 | |
| BC132207.1 | - | 2 - 3 | 136 | 40130484 | 62 | |
| BY083459.1 | - | 2 - 3 | 264 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anxa10
Genomic Map