Assay Details
| Gene Symbol: Anxa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136089.2 | NP_001129561.1 | 4 - 5 | 476 | 119 | |
| NM_011922.3 | NP_036052.2 | 4 - 5 | 476 | 119 | ||
| GenBank mRNA | AJ238978.1 | - | 4 - 5 | 376 | 119 | |
| AK008808.1 | - | 4 - 5 | 400 | 119 | ||
| AK018597.1 | - | 4 - 5 | 433 | 119 | ||
| AK020288.1 | - | 4 - 5 | 401 | 119 | ||
| AK131880.1 | - | 4 - 5 | 396 | 119 | ||
| AK168715.1 | - | 4 - 5 | 380 | 119 | ||
| BC125319.1 | - | 4 - 5 | 348 | 40129834 | 119 | |
| BC132207.1 | - | 4 - 5 | 348 | 40130484 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anxa10
Genomic Map