Assay Details
| Gene Symbol: Anxa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136089.2 | NP_001129561.1 | 9 - 10 | 885 | 159 | |
| NM_011922.3 | NP_036052.2 | 8 - 9 | 825 | 159 | ||
| GenBank mRNA | AK008772.1 | - | 9 - 10 | 808 | 159 | |
| AK008808.1 | - | 9 - 10 | 809 | 159 | ||
| AK020288.1 | - | 8 - 9 | 750 | 159 | ||
| AK131880.1 | - | 9 - 10 | 805 | 159 | ||
| AK168715.1 | - | 9 - 10 | 789 | 159 | ||
| BC125319.1 | - | 9 - 10 | 757 | 40129834 | 159 | |
| BC132207.1 | - | 9 - 10 | 757 | 40130484 | 159 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anxa10
Genomic Map