Assay Details
| Gene Symbol: Anxa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136089.2 | NP_001129561.1 | 6 - 7 | 648 | 76 | |
| GenBank mRNA | AJ238978.1 | - | 6 - 7 | 548 | 76 | |
| AK008772.1 | - | 6 - 7 | 571 | 76 | ||
| AK008808.1 | - | 6 - 7 | 572 | 76 | ||
| AK018597.1 | - | 6 - 7 | 605 | 76 | ||
| AK131880.1 | - | 6 - 7 | 568 | 76 | ||
| AK168715.1 | - | 6 - 7 | 552 | 76 | ||
| BC125319.1 | - | 6 - 7 | 520 | 40129834 | 76 | |
| BC132207.1 | - | 6 - 7 | 520 | 40130484 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anxa10
Genomic Map