Assay Details
| Gene Symbol: Nat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008674.2 | NP_032700.1 | 1 - 2 | 67 | 68 | |
| GenBank mRNA | AK077364.1 | - | 1 - 2 | 67 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077364.1, NM_008674.2 transcripts.
Set Membership: |
|

Nat3
Genomic Map