Assay Details
| Gene Symbol: Rfxank | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025589.1 | NP_001020760.1 | 4 - 5 | 622 | 63 | |
| GenBank mRNA | AK052298.1 | - | 4 - 5 | 579 | 63 | |
| AK146148.1 | - | 4 - 5 | 551 | 63 | ||
| BC010971.1 | - | 4 - 5 | 556 | 4190691 | 63 | |
More Information
Set Membership: |
|

Rfxank
Genomic Map