Assay Details
| Gene Symbol: Rfxank | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025589.1 | NP_001020760.1 | 5 - 6 | 722 | 82 | |
| NM_011266.2 | NP_035396.1 | 5 - 6 | 752 | 82 | ||
| GenBank mRNA | AF094761.1 | - | 5 - 6 | 668 | 82 | |
| AF123704.1 | - | 5 - 6 | 653 | 82 | ||
| AK052298.1 | - | 5 - 6 | 679 | 82 | ||
| AK087462.1 | - | 5 - 6 | 653 | 82 | ||
| AK146148.1 | - | 5 - 6 | 651 | 82 | ||
| AK162595.1 | - | 1 - 2 | 340 | 82 | ||
| BC010971.1 | - | 5 - 6 | 656 | 4190691 | 82 | |
| L43164.1 | - | 5 - 6 | 653 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfxank
Genomic Map