Assay Details
| Gene Symbol: Mef2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045484.1 | NP_001038949.1 | 7 - 8 | 799 | 88 | |
| NM_008578.2 | NP_032604.2 | 7 - 8 | 829 | 88 | ||
| GenBank mRNA | BC045147.1 | - | 7 - 8 | 799 | 6307224 | 88 |
| CO811773.1 | - | 466 | 88 | |||
| D87833.1 | - | 7 - 8 | 826 | 88 | ||
| D87835.1 | - | 7 - 8 | 796 | 88 | ||
More Information
Set Membership: |
|

Mef2b
Genomic Map