Assay Details
| Gene Symbol: 2310045N01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145552.1 | NP_001139024.1 | 2 - 3 | 179 | 72 | |
| GenBank mRNA | AK009829.1 | - | 2 - 3 | 168 | 72 | |
| BC048525.1 | - | 2 - 3 | 179 | 72 | ||
| BC065393.1 | - | 2 - 3 | 169 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

2310045N01Rik
Genomic Map