Assay Details
| Gene Symbol: Mef2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045484.1 | NP_001038949.1 | 2 - 3 | 143 | 98 | |
| NM_008578.2 | NP_032604.2 | 2 - 3 | 143 | 98 | ||
| GenBank mRNA | BC045147.1 | - | 2 - 3 | 143 | 6307224 | 98 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC045147.1, NM_001045484.1, NM_008578.2 transcripts.
Set Membership: |
|

Mef2b
Genomic Map