Assay Details
| Gene Symbol: Mef2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045484.1 | NP_001038949.1 | 3 - 4 | 342 | 73 | |
| NM_008578.2 | NP_032604.2 | 3 - 4 | 342 | 73 | ||
| GenBank mRNA | BB867153.1 | - | 3 - 4 | 370 | 74 | |
| BC045147.1 | - | 3 - 4 | 342 | 6307224 | 73 | |
More Information
Set Membership: |
|

Mef2b
Genomic Map