Assay Details
| Gene Symbol: Zfp866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177899.3 | NP_808567.2 | 3 - 4 | 307 | 118 | |
| GenBank mRNA | AK052363.1 | - | 3 - 4 | 286 | 118 | |
| AK133399.1 | - | 372 | 118 | |||
| AK137334.1 | - | 3 - 4 | 309 | 118 | ||
| AK140357.1 | - | 3 - 4 | 302 | 118 | ||
| BC057095.1 | - | 3 - 4 | 281 | 6419770 | 118 | |
| BC117846.1 | - | 3 - 4 | 262 | 40093653 | 118 | |
| BC117847.1 | - | 3 - 4 | 262 | 40093655 | 118 | |
| BC138224.1 | - | 3 - 4 | 258 | 8861243 | 118 | |
| BC138227.1 | - | 3 - 4 | 258 | 8861246 | 118 | |
More Information
Set Membership: |
|

Zfp866
Genomic Map