Assay Details
| Gene Symbol: Crtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004062.2 | NP_001004062.1 | 8 - 9 | 888 | 75 | |
| GenBank mRNA | AK129173.1 | - | 8 - 9 | 885 | 75 | |
| BC068139.1 | - | 8 - 9 | 894 | 75 | ||
| BC080308.1 | - | 8 - 9 | 888 | 5719562 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crtc1
Genomic Map