Assay Details
| Gene Symbol: Fkbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111066.1 | NP_001104536.1 | 4 - 5 | 639 | 94 | |
| NM_001199631.1 | NP_001186560.1 | 4 - 5 | 735 | 94 | ||
| GenBank mRNA | AK088739.1 | - | 4 - 5 | 574 | 94 | |
| AK152198.1 | - | 4 - 5 | 635 | 94 | ||
| AK153000.1 | - | 4 - 5 | 632 | 94 | ||
| AK158155.1 | - | 4 - 5 | 622 | 94 | ||
| AK165281.1 | - | 4 - 5 | 632 | 94 | ||
| BC003739.1 | - | 3 - 4 | 555 | 3601511 | 94 | |
| BC027808.1 | - | 4 - 5 | 589 | 5324380 | 94 | |
More Information
Set Membership: |
|

Fkbp8
Genomic Map