Assay Details
| Gene Symbol: Ssbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133772.2 | NP_598533.2 | 14 - 15 | 1060 | 53 | |
| GenBank mRNA | AK004835.1 | - | 14 - 15 | 1173 | 53 | |
| AK154336.1 | - | 14 - 15 | 1061 | 53 | ||
| AK157387.1 | - | 13 - 14 | 1140 | 53 | ||
| BC002150.1 | - | 14 - 15 | 1019 | 3484896 | 53 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ssbp4
Genomic Map