Assay Details
| Gene Symbol: Ssbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133772.2 | NP_598533.2 | 1 - 2 | 195 | 63 | |
| GenBank mRNA | AK154336.1 | - | 1 - 2 | 196 | 63 | |
| AK157387.1 | - | 1 - 2 | 184 | 63 | ||
| BC002150.1 | - | 1 - 2 | 154 | 3484896 | 63 | |
| CX214457.1 | - | 1 - 2 | 195 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ssbp4
Genomic Map