Assay Details
| Gene Symbol: Dda1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007085.1 | - | 1 - 2 | 152 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial ribosomal protein L34 | Mrpl34 |
0610007O17Rik, D8Bwg1484e |
-4.7kb | Sense | Chr. 8 - 71464926 - 71465753 on NCBI Build GRCm38 |
| Downstream | anoctamin 8 | Ano8 |
BC053460, Tmem16h |
3.9kb | Anti-sense | Chr. 8 - 71476017 - 71486067 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Dda1
Genomic Map