Assay Details
| Gene Symbol: Cd72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | J04170.1 | - | 5 - 6 | 724 | 108 | |
| S40777.1 | - | 5 - 6 | 720 | 108 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Cd72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | J04170.1 | - | 5 - 6 | 724 | 108 | |
| S40777.1 | - | 5 - 6 | 720 | 108 | ||
Set Membership: |
|