Assay Details
| Gene Symbol: Arl13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026577.3 | NP_080853.3 | 2 - 3 | 426 | 63 | |
| GenBank mRNA | AK016794.1 | - | 2 - 3 | 372 | 63 | |
| AK020859.1 | - | 2 - 3 | 367 | 63 | ||
| AK051159.1 | - | 2 - 3 | 384 | 63 | ||
| BC082574.1 | - | 2 - 3 | 426 | 30550373 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arl13b
Genomic Map