Assay Details
| Gene Symbol: Arl13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026577.3 | NP_080853.3 | 3 - 4 | 681 | 82 | |
| GenBank mRNA | AK016794.1 | - | 3 - 4 | 627 | 82 | |
| AK021270.1 | - | 3 - 4 | 683 | 82 | ||
| BC082574.1 | - | 3 - 4 | 681 | 30550373 | 82 | |
More Information
Set Membership: |
|

Arl13b
Genomic Map