Assay Details
| Gene Symbol: Mef2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170537.1 | NP_001164008.1 | 10 - 11 | 1650 | 88 | |
| GenBank mRNA | AK132573.1 | - | 10 - 11 | 1479 | 88 | |
| BC037731.1 | - | 10 - 11 | 1396 | 4500786 | 88 | |
| BC057650.1 | - | 10 - 11 | 1385 | 4500786 | 88 | |
| L13171.1 | - | 8 - 9 | 1211 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mef2c
Genomic Map