| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300015077.mm.1 |
NM_001040692.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015077.mm.1 |
NM_001040692.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015077.mm.1 |
NM_001136087.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015077.mm.1 |
NM_001136087.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015077.mm.1 |
NM_001168643.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015077.mm.1 |
NM_001168643.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015077.mm.1 |
NM_001168644.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015077.mm.1 |
NM_001168644.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015077.mm.1 |
NM_001168645.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015077.mm.1 |
NM_001168645.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015077.mm.1 |
NM_001168646.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015077.mm.1 |
NM_001168646.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015077.mm.1 |
TC1300002302.mm.1 |
MTA TC |